Learn More
Macherey-Nagel™ Alugram Aluminum Sheet Nano-SILGUR
Brand:  Macherey-Nagel™ 818432
Product Code. 12710853
  • £198.00 / Pack of 25
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Specifications
2 to 10 μm
25 Pk.
0.75 mL/g
0.02 mm
60 Å
Aluminum Sheet
∼500 m2/g
10 x 10 cm