missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ ALUGRAM™ Xtra SIL G Aluminum Sheets

Product Code. 12720743 Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 7.5 cm
5 x 10 cm
5 x 20 cm
20 x 20 cm
Unit Size:
Pack of 20
Pack of 25
Pack of 50
This item is not returnable. View return policy

Product Code. 12720743

Brand: Macherey-Nagel™ 818230.20

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Stable in almost all organic solvents and in purely aqueous eluents. Macherey-Nagel™ ALUGRAM™ Xtra SIL G Aluminum Sheets are unmodified standard silica layers on aluminum for use in TLC. Feature outstanding wettability and improved cutting properties.

ALUGRAM™ Xtra SIL G Aluminum Sheets feature unmodified, standard-grade silica 60 layers without a fluorescent indicator.

  • Outstanding wettability for precise colorization results, even with 100% aqueous detection reagents
  • Excellent separation efficiency and reproducibility from lot to lot
  • Easy and reliable cutting due to an optimized binder system, no flaking of silica
  • Stable in almost all organic solvents and resistant towards aggressive visualization reagents
  • Highly polymeric product is also completely stable in purely aqueous eluents

Specifications

Material Alugram Aluminum Sheets with IUInmodified Silica 60 Layers
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 20 Pk.
Type Aluminum Sheet
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.20 mm
Dimensions (L x W) 5 x 7.5 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.