missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ ALUGRAM™ Xtra SIL G Aluminum Sheets
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 7.5 cm
5 x 10 cm
5 x 20 cm
20 x 20 cm
Unit Size:
Pack of 20
Pack of 25
Pack of 50
Beschreibung
ALUGRAM™ Xtra SIL G Aluminum Sheets feature unmodified, standard-grade silica 60 layers without a fluorescent indicator.
- Outstanding wettability for precise colorization results, even with 100% aqueous detection reagents
- Excellent separation efficiency and reproducibility from lot to lot
- Easy and reliable cutting due to an optimized binder system, no flaking of silica
- Stable in almost all organic solvents and resistant towards aggressive visualization reagents
- Highly polymeric product is also completely stable in purely aqueous eluents
Spezifikation
Spezifikation
| Material | Alugram Aluminum Sheets with IUInmodified Silica 60 Layers |
| Particle Size | 5 to 17 μm |
| Pore Size | 60 Å |
| Quantity | 50 Pk. |
| Type | Aluminum Sheet |
| Pore Volume | 0.75 mL/g |
| Specific Surface | ∼500 m2/g |
| Thickness (Metric) | 0.20 mm |
| Dimensions (L x W) | 5 x 20 cm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur