missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.

Brand:  Macherey-Nagel™ 818332

74.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12730773

  • £139.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

 
  • Unmodified standard silica layers on aluminum for TLC
  • Silica 60
  • Outstanding wettability for precise colorization results, even with 100 % aqueous detection reagents
  • Excellent separation efficiency and reproducibility from lot to lot
  • Easy and reliable cutting due to an optimized binder system, no flaking of silica
  • Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents; also completely stable in purely aqueous eluents
Specifications

Specifications

5 to 17 μm
50 Pk.
UV254
∼500 m2/g
5 x 20 cm
60 Å
Aluminum Sheet
0.75 mL/g
0.20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.