missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254

Product Code. 12804281 Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
4 x 8 cm
5 x 7.5 cm
5 x 10 cm
5 x 20 cm
10 x 20 cm
20 x 20 cm
Unit Size:
Pack of 20
Pack of 25
Pack of 50
This item is not returnable. View return policy

Product Code. 12804281

Brand: Macherey-Nagel™ 818333

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.

  • Unmodified standard silica layers on aluminum for TLC
  • Silica 60
  • Outstanding wettability for precise colorization results, even with 100 % aqueous detection reagents
  • Excellent separation efficiency and reproducibility from lot to lot
  • Easy and reliable cutting due to an optimized binder system, no flaking of silica
  • Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents; also completely stable in purely aqueous eluents

Specifications

Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 25 Pk.
Type Aluminum Sheet
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.20 mm
Dimensions (L x W) 20 x 20 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.