Learn More
Aluminum chloride, 98.5%, extra pure, anhydrous, powder, Thermo Scientific Chemicals
£20.56 - £75.10
Chemical Identifiers
CAS | 7446-70-0 |
---|---|
Molecular Formula | AlCl3 |
Molecular Weight (g/mol) | 133.33 |
MDL Number | MFCD00003422 |
InChI Key | VSCWAEJMTAWNJL-UHFFFAOYSA-K |
Synonym | aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminumShow More |
IUPAC Name | aluminum(3+) trichloride |
SMILES | [Al+3].[Cl-].[Cl-].[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10165080
|
Thermo Scientific Acros
217460050 |
5 g | Glass bottle |
£20.56
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10358540
|
Thermo Scientific Acros
217461000 |
100 g | Glass bottle |
£24.25
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10266700
|
Thermo Scientific Acros
217465000 |
500 g | Glass bottle |
£31.35
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10065363
|
Thermo Scientific Acros
217460025 |
2.5 kg | Glass bottle | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Lewis acidChemical Identifiers
7446-70-0 | |
133.33 | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride |
AlCl3 | |
MFCD00003422 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminumShow More | |
[Al+3].[Cl-].[Cl-].[Cl-] |
Specifications
194.0°C | |
Gray or White to Yellow | |
98% min. (Argentometry) | |
AlCl3 | |
01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride | |
133.34 | |
Extra Pure | |
2.4400g/mL |
7446-70-0 | |
Powder | |
AlCl3 | |
MFCD00003422 | |
15, 333 | |
Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform | |
[Al+3].[Cl-].[Cl-].[Cl-] | |
133.33 | |
98.5% | |
2.44 | |
Aluminum chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-208-1
missing translation for 'rtecsNumber' : BD0525000
missing translation for 'tsca' : TSCA
- Aluminium chloride
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.