Learn More
Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucible
Use these crucibles for general laboratory applications. Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucibles are resistant to a variety of acids and alkalis.
Brand:  Haldenwanger™ L ALSINTTIEGEL*B/2
 View more versions of this product
Product Code. 10291971
  • £276.00 / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Conical squat form crucibles
  • Aluminum ceramic composition
  • Available in a variety of sizes
  • Aluminum ceramic composition
Specifications
Specifications
Aluminum Ceramic
60 mL
Crucible
Conical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucible > Capacity: 60mL