Learn More
Invitrogen™ FluoCells™ Prepared Slide #2 (BPAE cells with Mouse Anti-α-tubulin, AlexaFluor™ 488, FL Goat Anti-Mouse IgG, Texas Red™-X Phalloidin, and DAPI)
Multilabeled cell preparations for observation by fluorescence microscopy
Brand: Invitrogen™ F14781
202.16 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
FluoCells prepared slide #2 contains bovine pulmonary artery endothelial cells (BPAEC). Texas Red-X phalloidin is labeling the F-actin, anti—bovine α-tubulin mouse monoclonal 236-10501 in conjunction with BODIPY FL goat anti—mouse IgG antibody is labeling microtubules, and DAPI is labeling the nucleus.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF)
Order Info
Shipping Condition: Room temperature

Specifications
Prepared Microscope Slide | |
Immunofluorescence | |
1 Each |
BODIPY FluoCells Texas Red | |
Store at room temperature and protect from light. | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.