missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DMEM, high glucose

DMEM, high glucose
Brand: Gibco™ 41965039
10.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications.DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM product made in our Grand Island facility (11965-084). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Specifications
1 X | |
Dulbecco's Modified Eagle Medium | |
No HEPES, No Sodium Pyruvate | |
500 mL | |
7.0 to 7.4 | |
Dulbecco's Modified Eagle Medium (DMEM), 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With High Glucose, L-Glutamine, Phenol Red, 12 Months Shelf Life, 320 to 360mOsm/kg Osmolality, For Mammalian Cell Culture |
Liquid | |
High Glucose, Glutamine, Phenol Red | |
Tissue and cell culture processing, mammalian cell culture | |
ISO 13485 standards, FDA | |
2 to 8°C |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ DMEM, high glucose > 500mL
Spot an opportunity for improvement?Share a Content Correction