Learn More
Gibco™ Fetal Bovine Serum, Value
Gibco fetal bovine sera offers an excellent value for basic cell culture, specialty research, and specific assays. It has earned the trust of researchers with consistent quality and award-winning support that helps meet research needs and budget requirements.
Brand: Gibco™ A5256701
248.25 GBP valid until 2025-12-31
Use promo code "24071" to get your promotional price.
Includes: Gibco A5256701
Description
Gibco Serum Delivers
- ISO 13485-certified, processed in FDA-registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle is easier to use in hood, reduces risk of contamination, and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
All products may not be available in all regions due to importation regulations
US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that help you meet your research needs and budget requirements.
- Sera category: Value
- Sera application: Cell culture with robust cell lines, good value for basic research
- Testing: Up to 30 quality tests
- Endotoxin level: ≤20 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
- Origin: Refer to CoA for specific origin

Specifications
Store at <-10°C. | |
Cattle/Bovine | |
Refer to CoA for specific origin | |
Fetal Bovine Serum | |
Sterile | |
Up to 30 quality tests including 9CFR virus testing* endotoxin, performance (*Please note: If manufactured in the UK, Full 9CFR except for Rabies and BlueTongue Virus which is tested via EMA) | |
Less waste | |
≤25 mg/dL | |
Dry Ice | |
No |
Gibco A5256701 | |
Fetal | |
Bottle | |
500 mL | |
Triple-filtered, 0.1 μm | |
≤20 EU/mL | |
No | |
Value | |
USDA Safety Testing Performed for Mexican and Australian origins |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2529135 | Certificate of Analysis | 29/01/2025 | A5256701 |
2552471 | Certificate of Analysis | 29/01/2025 | A5256701 |
2503996 | Certificate of Analysis | 29/01/2025 | A5256701 |
2529136 | Certificate of Analysis | 29/01/2025 | A5256701 |
2563339 | Certificate of Analysis | 29/01/2025 | A5256701 |
Special Offers
-
Thermo Scientific Nunc
,
Thermo Scientific Nalgene
,
Gibco
,
Invitrogen
,
Thermo Scientific
Cell Biology Promotion
Discover how you can maximise your savings and boost your research to the next level with our top-of-the-line Gibco and Thermo Scientific products.Promo Code: 24071Expires: 2025-06-30
Days
66Hours
4Minutes
56Seconds
34
Your input is important to us. Please complete this form to provide feedback related to the content on this product.