Learn More
Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucible
Use these crucibles for general laboratory applications. Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucibles are resistant to a variety of acids and alkalis.
Brand:  Haldenwanger™ L ALSINTTIEGEL*B/00
 View more versions of this product
Product Code. 10757591
  • £38.15 / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Conical squat form crucibles
  • Aluminum ceramic composition
  • Available in a variety of sizes
  • Aluminum ceramic composition
Specifications
Specifications
Aluminum Ceramic
10 mL
Crucible
Conical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucible > Capacity: 10mL