Learn More
Aluminum chloride, anhydrous, Reagent Grade, Thermo Scientific Chemicals
£36.47 - £275.37
Chemical Identifiers
CAS | 7446-70-0 |
---|---|
Molecular Formula | AlCl3 |
Molecular Weight (g/mol) | 133.33 |
MDL Number | MFCD00003422 |
InChI Key | VSCWAEJMTAWNJL-UHFFFAOYSA-K |
Synonym | aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminumShow More |
IUPAC Name | aluminum(3+) trichloride |
SMILES | [Al+3].[Cl-].[Cl-].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381649
|
Thermo Scientific Alfa Aesar
012298.30 |
250 g |
£36.47
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371649
|
Thermo Scientific Alfa Aesar
012298.A1 |
1 kg |
£57.62
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361649
|
Thermo Scientific Alfa Aesar
012298.D9 |
5 x 1 kg |
£275.37
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a catalyst for Friedel-Crafts acylation and alkylation of aromatic compounds. It is one of the most commonly employed Lewis acids for a wide variety of organic transformations. It catalyzes the ene reaction, polymerization, and isomerization reactions. For example, it can be used to synthesis ethyl benzene which is a precursor for producing polystyrene. It can be used for producing dodecylbenzene, a key intermediate for detergents. It is useful in the production of anthroquinone, the presursor for dyestuffs. It is used in the synthesis of bis(arene) metal complexes, through Fischer-Hafner synthesis. Gattermann-Koch reaction employs aluminum chloride for introducing formyl group onto aromatic rings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7446-70-0 | |
133.33 | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride |
AlCl3 | |
MFCD00003422 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminumShow More | |
[Al+3].[Cl-].[Cl-].[Cl-] |
Specifications
190° (sublimation) | |
Gray to Yellow | |
AlCl3 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride | |
133.34 | |
Pungent | |
2.44 |
7446-70-0 | |
Lump/Powder | |
MFCD00003422 | |
Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile. | |
[Al+3].[Cl-].[Cl-].[Cl-] | |
133.33 | |
Reagent | |
Moisture sensitive | |
Aluminum chloride |
Safety and Handling
Keep container tightly closed and dry. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II
missing translation for 'einecsNumber' : 231-208-1
missing translation for 'rtecsNumber' : BD0525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum chloride, anhydrous
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.