missing translation for 'onlineSavingsMsg'
Learn More
Aluminum chloride, anhydrous, Reagent Grade, Thermo Scientific Chemicals Product Code.: 11361649

Aluminum chloride, anhydrous, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11361649
5 x 1 kg
Click to view available options
Quantity:
250 g
1 kg
5 x 1 kg
This item is not returnable. View return policy

Product Code. 11361649

Brand: Thermo Scientific Alfa Aesar 012298.D9

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a catalyst for Friedel-Crafts acylation and alkylation of aromatic compounds. It is one of the most commonly employed Lewis acids for a wide variety of organic transformations. It catalyzes the ene reaction, polymerization, and isomerization reactions. For example, it can be used to synthesis ethyl benzene which is a precursor for producing polystyrene. It can be used for producing dodecylbenzene, a key intermediate for detergents. It is useful in the production of anthroquinone, the presursor for dyestuffs. It is used in the synthesis of bis(arene) metal complexes, through Fischer-Hafner synthesis. Gattermann-Koch reaction employs aluminum chloride for introducing formyl group onto aromatic rings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminumShow MoreShow Less
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 190° (sublimation)
Color Gray to Yellow
Physical Form Lump/Powder
Quantity 5 x 1 kg
Solubility Information Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile.
Formula Weight 133.34
Grade Reagent
Odor Pungent
Sensitivity Moisture sensitive
Density 2.44
Chemical Name or Material Aluminum chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX30K036 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number012298.D9,012298.A1,012298.30
Lot NumberS27K025 Certificate TypeCertificate of Analysis Date17/07/2024 Catalog Number012298.D9,012298.A1,012298.30
Lot NumberR17K033 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number012298.D9,012298.A1,012298.30
Lot NumberM31K027 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog Number012298.D9,012298.A1,012298.30
Lot NumberZ28J029 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number012298.D9,012298.A1,012298.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum chloride, anhydrous, Reagent Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.