missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Cobalt powder, -100 mesh, 99% (metals basis), Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 088320.36

695.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11329249

  • £698.48 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aluminum Cobalt powder
(metals basis)
UN3132
99%
Odorless
Al:Co; 69:31 wt%
MFCD00239520
500 g
−100 Mesh
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW25K011 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number088320.36
Lot NumberL23N39 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number088320.36
Lot NumberK06Z028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088320.36
Lot NumberX16E027 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088320.36
Lot NumberH08K11 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088320.36
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum Cobalt powder, -100 mesh, 99% (metals basis), Thermo Scientific Chemicals > 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only