Learn More
Boron tribromide, 1M solution in methylene chloride, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 198900100
24.05 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
35.90 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acid
Specifications
Boron tribromide | |
10294-33-4, 75-09-2 | |
Solution | |
BBr3 | |
MFCD00011312 | |
15, 1349 | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in alcohol and ccl4 | |
BrB(Br)Br | |
250.52 | |
250.52 | |
Glass bottle | |
1.4670g/mL |
1M Solution in Methylene Chloride | |
Yellow to Brown | |
10 mL | |
BBr3 | |
01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43 | |
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
tribromoborane | |
25134 | |
0.9 to 1.1M | |
1.467 |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Boron tribromide, 1M solution in methylene chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H300-Fatal if swallowed.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468183 | Certificate of Analysis | 03/12/2024 | 198900100,198908000,198901000 |
A0463732 | Certificate of Analysis | 12/06/2024 | 198900100,198908000,198901000 |
A0463733 | Certificate of Analysis | 12/06/2024 | 198900100,198908000,198901000 |
A0456186 | Certificate of Analysis | 08/09/2023 | 198900100,198908000,198901000 |
A0456187 | Certificate of Analysis | 07/09/2023 | 198900100,198908000,198901000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only