Learn More
Boron tribromide, 1M solution in methylene chloride, Thermo Scientific Chemicals
£36.08 - £539.69
Specifications
Chemical Name or Material | Boron tribromide |
---|---|
Name Note | 1M Solution in Methylene Chloride |
CAS | 10294-33-4, 75-09-2 |
Color | Yellow to Brown |
Physical Form | Solution |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10398190
|
Thermo Scientific Acros
198900100 |
10 mL | Glass bottle |
£36.08
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10552221
|
Thermo Scientific Acros
198901000 |
100 mL | Glass bottle |
£112.56
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10733894
|
Thermo Scientific
198908000 |
800 mL | Glass bottle |
£539.69
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acidSpecifications
Boron tribromide | |
10294-33-4, 75-09-2 | |
Solution | |
BBr3 | |
01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43 | |
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
BrB(Br)Br | |
250.52 | |
250.52 | |
Glass bottle | |
1.4670g/mL |
1M Solution in Methylene Chloride | |
Yellow to Brown | |
BBr3 | |
MFCD00011312 | |
15, 1349 | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
tribromoborane | |
25134 | |
0.9 to 1.1M | |
1.467 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Fatal if swallowed.
Suspected of causing cancer.
Reacts violently with water.
May cause drowsiness or dizziness.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-657-9
Recommended Storage : Darkens on storage
- Boron tribromide, 1M solution in methylene chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H300-Fatal if swallowed.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.