missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Aluminum Crimp Seals
Seals freeze drying and injection vials. Azpack™Aluminum Crimp Seals prevent leakage of hazardous and biological products.
Brand: Azpack™ 10294854
Description
- For use with clear neutral glass vials
- Aluminum
- 500/Pack
Specifications
Silver | |
Aluminum | |
Crimp Cap |
13 mm | |
2 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction