missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Aluminum Crimp Seals
Seals freeze drying and injection vials. Azpack™Aluminum Crimp Seals prevent leakage of hazardous and biological products.
£104.00 - £115.00
Product Code | Brand | Diameter (Metric) Outer | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Volume (Metric) | Price | Quantity & Availability | ||||
11318133
|
Azpack™
11318133 |
20 mm | 5, 10, 20 mL |
£104.00
Pack of 500 |
In Stock
Log in to see stock. |
||||
10294854
|
Azpack™
10294854 |
13 mm | 2 mL |
£115.00
Pack of 500 |
In Stock
Log in to see stock. |
||||
Description
- For use with clear neutral glass vials
- Aluminum
- 500/Pack
Specifications
Silver | |
Aluminum | |
Crimp Cap |