missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Aluminum Horizontal Freezer Rack

Wesbart™ Aluminum Horizontal Freezer Racks are front loading with trays for quick access to cryoboxes and other stored items.

Brand:  Wesbart™ AH179/3UT

Code : NEW

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 15726418

  • £123.00 / Each
Estimated Shipment: 10-10-2024
to see stock.
Add to basket

Description

Description

The main benefit of the front access rack is to gain access to the samples only one tray has to be removed from the rack meaning all other samples are kept at low temperature.
Specifications

Specifications

Boxes (12/Rack)
3 Trays/Rack
Suitable for 4 in. cryoboxes
Aluminum
3
560 x 140 x 420 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers