Learn More
Wesbart™ Aluminum Horizontal Freezer Rack
Wesbart™ Aluminum Horizontal Freezer Racks are front loading with trays for quick access to cryoboxes and other stored items.
Brand: Wesbart™
Additional Details : Weight : 2.50000kg
Description
- Aluminum front access storage rack for tray units (ordered separately, Cat. No. 17311849) for upright freezers
- For standard cryobox or other freezer storage
- Only one tray has to be removed from the rack at a time for sample access, minimizing temperature impact on remaining items
- Range of sizes and capacities available
Specifications
Freezer Storage Rack | |
Aluminum | |
Upright Freezers |
Loose Items or Cryoboxes | |
1 Tray/Rack | |
560 x 140 x 290 mm |