missing translation for 'onlineSavingsMsg'
Learn More

Aluminum hydroxide, 76.5% min, Thermo Scientific Chemicals

£22.71 - £119.60

Chemical Identifiers

CAS 21645-51-2
Molecular Formula AlH3O3
Molecular Weight (g/mol) 78.00
MDL Number MFCD00003420
InChI Key WNROFYMDJYEPJX-UHFFFAOYSA-K
IUPAC Name aluminium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Al+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311759
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012366.22
100 g
£22.71
100g
Please to purchase this item. Need a web account? Register with us today!
11301759
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012366.36
500 g
£71.46
500g
Please to purchase this item. Need a web account? Register with us today!
11321759
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012366.A3
2 kg
£119.60
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmeticsAluminum hydroxide is used as a desiccant powder, filler in paper, plastics, rubber and cosmetics. It is used as a smoke suppressant and mordant dye. It is also used in drugs as antacid and antihyperphosphatemic. It is also used as a Claus catalyst support for waterproof fabrics. It is an important starting material for the preparation of other aluminum compounds, calcined alumina, aluminum sulfate, polyaluminum chloride, zeolites, sodium aluminate, activated alumina and aluminum nitrate. In addition, it is used as a fire retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21645-51-2
78.00
WNROFYMDJYEPJX-UHFFFAOYSA-K
[OH-].[OH-].[OH-].[Al+3]
AlH3O3
MFCD00003420
aluminium(3+) trihydroxide

Specifications

21645-51-2
76.5% min.
MFCD00003420
WNROFYMDJYEPJX-UHFFFAOYSA-K
aluminium(3+) trihydroxide
78
Aluminum hydroxide
Granules
AlH3O3
Soluble in alkaline aqueous solution,hydrochloric acid and sulfuric acid solution. Insoluble in water.
[OH-].[OH-].[OH-].[Al+3]
78.00
Reagent
Videos
Safety and Handling

Safety and Handling

H315-H319-H335

missing translation for 'einecsNumber' : 244-492-7

missing translation for 'rtecsNumber' : BD0940000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Aluminum hydroxide

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only