missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments Aluminum Oil Bath for MR300X

Aluminum Oil Bath for MR300X with 2 or 4 liter capacity for maximum temperature of 250°C that is ideal for use in research, scientific, and laboratory environments.

Brand:  Heidolph Instruments 504-92000-00

 View more versions of this product

Product Code. 10334621

  • £359.00 / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Aluminum bath for superior heat transfer
  • Comes with two insulated safety handles and safety rim for secure mounting on the hot plate
  • Heidolph oil bath for max. temp. of 250°C
  • Provide more uniform heat in comparison to other heating devices
  • Ideal for use in research, scientific, and laboratory environment
Specifications

Specifications

2 L
482°F
Aluminum
188 x 121 mm
250°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.