missing translation for 'onlineSavingsMsg'
Learn More
Aluminum oxide, calcined, insulating powder, Thermo Scientific Chemicals Product Code.: 11363128

Aluminum oxide, calcined, insulating powder, Thermo Scientific Chemicals

Product Code. 11363128
1 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11363128

Brand: Thermo Scientific Alfa Aesar 033195.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Quantity 1 kg
IUPAC Name dialuminum;oxygen(2-)
Odor Odorless
Chemical Name or Material Aluminum oxide, Calcined, Insulating powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN24L042 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog Number033195.A1,033195.A7
Lot NumberP08K012 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog Number033195.A1
Lot NumberM24J004 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number033195.A1,033195.A7
Lot NumberT19J006 Certificate TypeCertificate of Analysis Date25/07/2023 Catalog Number033195.A1,033195.A7
Lot NumberN17J053 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number033195.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum oxide, calcined, insulating powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.