Learn More
Haldenwanger™ Aluminum Oxide Combustion Boats
Use these boats for a variety laboratory applications. Haldenwanger™ Aluminum Oxide Combustion Boats are ideal for chemical calcining, metal casting and sample preparation.
Brand:  Haldenwanger™ L ALS.SCH.75X11X7
 View more versions of this product
Product Code. 12317018
  • £132.00 / Each
Explore more special offers
Description
Description
Aluminum oxide boats of great hardness, strength and chemical resistance. Suitable for continuous use up to 1, 700°C to 1, 750°C.
Specifications
Specifications
Combustion Boat
7 mm
1 Ea.
For continuous use up to 1700 to 1750°C.
Aluminum Oxide
75 mm