missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Aluminum Oxide Combustion Boats

Use these boats for a variety laboratory applications. Haldenwanger™ Aluminum Oxide Combustion Boats are ideal for chemical calcining, metal casting and sample preparation.

Brand:  Haldenwanger™ L ALS.SCH.160X40X21

 View more versions of this product

Product Code. 12357018

  • £394.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Aluminum oxide boats
  • Great hardness and strength
  • Resistant to heat and caustic chemicals
  • Suitable for continuous use up to 1,750°C
Specifications

Specifications

Combustion Boat
21 mm
1 Ea.
For continuous use up to 1700 to 1750°C.
Aluminum Oxide
160 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.