missing translation for 'onlineSavingsMsg'
Learn More
Aluminum oxide, gamma-phase, 99.97% (metals basis), Thermo Scientific Chemicals Product Code.: 11389117

Aluminum oxide, gamma-phase, 99.97% (metals basis), Thermo Scientific Chemicals

Product Code. 11389117
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11389117

Brand: Thermo Scientific Alfa Aesar 039812.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Aluminum oxide, is useful to produce aluminum metal. It has many uses in pharmaceutical and industrial manufacturing processes as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements, available in toothpaste, as a dispersing agent food additive, and for uses in hemodialysis. Tailored aluminum oxide layers plays an important role for the bipolar current adjustment in the plasma electrolytic oxidation process. Red phosphorus/aluminum oxide composites play an essential role as flame retardants in recycled poly(ethylene terephthalate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.97% (Metals basis)
Solubility Information Slightly soluble in cold water,acid and alkali. Insoluble in hot water.
Formula Weight 101.96
Odor Odorless
Chemical Name or Material Aluminum oxide, gamma

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU13K007 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number039812.14,039812.36,039812.22
Lot NumberU13K006 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number039812.14,039812.36,039812.22
Lot NumberU10J002 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number039812.14,039812.36,039812.22
Lot NumberU16J002 Certificate TypeCertificate of Analysis Date06/09/2023 Catalog Number039812.14,039812.36,039812.22
Lot NumberI03Z005 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number039812.14,039812.36,039812.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum oxide, gamma-phase, 99.97% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.