missing translation for 'onlineSavingsMsg'
Learn More
Aluminum oxide, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals Product Code.: 10042690

Aluminum oxide, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Product Code. 10042690
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10042690

Brand: Thermo Scientific Acros 212250250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2977.0°C
Melting Point 2030.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99%
Linear Formula AI2O3
Fieser 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9
Merck Index 15, 350
Solubility Information Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic, solvents, difficultly soluble in mineral acids
Formula Weight 101.96
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 3.97
Density 3.9700g/mL
Chemical Name or Material Aluminum oxide, Extra Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466545 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number212250250,212251000
Lot NumberA0462478 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog Number212250250,212251000
Lot NumberA0454718 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number212250250,212251000
Lot NumberA0398944 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number212250250,212251000
Lot NumberA0405082 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number212250250,212251000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum oxide, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.