missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Standard SIL G Silica Layers on Alugram™ Aluminum Sheets
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets come in a variety of sizes.
£22.15 - £234.00
Specifications
Particle Size | 5 to 17 μm |
---|---|
Pore Size | 60 Å |
Type | TLC Plate |
For Use With (Application) | Thin Layer Chromatography (TLC) |
Pore Volume | 0.75 mL/g |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10519832
|
Macherey-Nagel™
818033 |
20 x 20 cm |
£234.00
Pack of 25 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
10786902
|
Macherey-Nagel™
818032 |
5 x 20 cm |
£80.20
Pack of 50 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
10033031
|
Macherey-Nagel™
818030.20 |
5 x 7.5 cm |
£22.15
Pack of 20 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
10780323
|
Macherey-Nagel™
818163 |
10 x 20 cm |
£110.00
Pack of 20 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
10377931
|
Macherey-Nagel™
818161 |
5 x 10 cm |
£52.20
Pack of 50 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets come in a variety of sizes.
Alugram™ Aluminum Sheets for TLC- Pre-coated ready to use plates
- Broad range of plate sizes, surface coatings and backings
- Silica 60
- specific surface (BET) ∼ 500m2/g
- Mean pore size 60 Å
- Specific pore volume 0.75mL/g,
- Particle size 5–17μm
- Standard grade
- For analytical plates 0.25mm
- For preparative plates 0.5 and 1mm
- For 2mm preparative layers, a slightly coarser material is used
- Manganese activated zinc silicate with green fluorescence for short-wave UV (254nm)
- Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366nm)
- Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications
5 to 17 μm | |
TLC Plate | |
0.75 mL/g | |
0.2 mm |
60 Å | |
Thin Layer Chromatography (TLC) | |
∼500 m2/g |