Learn More
Macherey-Nagel™ Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.
Brand: Macherey-Nagel™ 818130.20
23.51 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.
Alugram™ Aluminum Sheets for TLC- Pre-coated ready to use plates
- Broad range of plate sizes, surface coatings and backings
- Silica 60
- specific surface (BET) ∼ 500 m2/g
- Mean pore size 60 Å
- Specific pore volume 0.75 mL/g,
- Particle size 5–17 μm
- Standard grade
- For analytical plates 0.25 mm
- For preparative plates 0.5 and 1 mm
- For 2 mm preparative layers a slightly coarser material is used.
- Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
- Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
- Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications
Aluminum Sheets Alugram SIL G | |
60 Å | |
TLC Plate | |
0.75 mL/g | |
0.2 mm |
5 to 17 μm | |
20 Pk. | |
UV254 | |
∼500 m2/g | |
5 x 7.5 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.