Learn More
AM966, MedChemExpress
MedChemExpress AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).
Brand:  Medchem Express HY-15277
Encompass_Preferred
 View more versions of this product
Product Code. 16456418
  • £425.92 / 10mg
Explore more special offers
Description
Description
AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).
Specifications
Specifications
AM966
Off-White
C27H23ClN2O5
DMSO : 100 mg/mL (203.70 mM; Need ultrasonic)
490.93
98.64%
Solid
1228690-19-4
Research
10 mg
O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4Cl)C)=O)C(C)=NO3)C=C2)C=C1
490.93
Research
COVID-19-immunoregulation