missing translation for 'onlineSavingsMsg'
Learn More

AM966, MedChemExpress

MedChemExpress AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).

£246.84 - £1510.08

Specifications

Chemical Name or Material AM966
CAS 1228690-19-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H23ClN2O5
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16476706
Encompass_Preferred
View Documents
Medchem Express
HY-15277
100 mg
£1510.08
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16456418
Encompass_Preferred
View Documents
Medchem Express
HY-15277
10 mg
£425.92
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16439007
Encompass_Preferred
View Documents
Medchem Express
HY-15277
5 mg
£246.84
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16422527
Encompass_Preferred
View Documents
Medchem Express
HY-15277
50 mg
£1235.41
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).
Specifications

Specifications

AM966
Off-White
C27H23ClN2O5
O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4Cl)C)=O)C(C)=NO3)C=C2)C=C1
490.93
Research
COVID-19-immunoregulation
1228690-19-4
Research
DMSO : 100 mg/mL (203.70 mM; Need ultrasonic)
490.93
98.64%
Solid
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.