Learn More
BRAND™ Amber Conical Ground Joint Stoppers
Boro 3.3 honed stoppers
Brand:  BRAND™ 144672
6.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15670479
  • £6.80 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Honed conical joint stopper
1
NS 12/21
Borosilicate Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    22
    Hours
    10
    Minutes
    2
    Seconds
    21
Product Content Correction
Product Title
BRAND™ Amber Conical Ground Joint Stoppers > Size: NS 12/21