Learn More
Invitrogen™ CTP Soln. (10 mM)
This Invitrogen solution is a component of the MAXIscript™ Kits and is supplied in one tube containing 100 μL.
Brand: Invitrogen™ AM8120G
Description
This Invitrogen solution is a component of the MAXIscript™ Kits (Cat. No. AM1312, AM1316, AM1314, AM1326, AM1324, AM1318, AM1322, AM1320, AM1310, and AM1308) and is supplied in one tube containing 100 μL. Each is subjected to RNase testing as well as a functional assay to ensure high levels of incorporation.
Order Info
Shipping Condition: Dry Ice

Specifications
Store at -20°C. | |
In Vitro transcription | |
Ambion™ | |
Dry Ice |
10 mM | |
Unlabeled | |
100 μL |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3058732 | Certificate of Analysis | 25/01/2025 | AM8120G |
2735650 | Certificate of Analysis | 14/08/2023 | AM8120G |
2723347 | Certificate of Analysis | 16/06/2023 | AM8120G |
2644907 | Certificate of Analysis | 31/03/2023 | AM8120G |
2631456 | Certificate of Analysis | 30/01/2023 | AM8120G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.