Learn More
Invitrogen™ GLOBINclear™-Human Kit, for globin mRNA depletion
A rapid and robust treatment for RNA derived from whole blood that uses a novel, non-enzymatic technology to rapidly deplete >95% of the alpha and beta globin mRNA from total RNA preparations.
Brand: Invitrogen™ AM1980
Includes: 20μL Capture Oligo Mix; 600μL Streptavidin Magnetic Beads; 1.5mL 2X Hybridization Buffer; 1.5mL Streptavidin Bead Buffer; 1.75 Nuclease-free Water; 200μL RNA Binding Beads; 5mL RNA Wash Solution Concentrate; 1mL Elution Buffer; 80μL RNA Bead Buffer; 4mL RNA Binding Buffer Concentrate; 100 1.5-ml Non-stick Tubes
Requires : Kit requires use of a magnetic stand. The Six-Tube Magnetic Stand (Cat. No. AM10055) can hold six separate 1.5mL conical microcentrifuge tubes or 2mL round-bottomed microtubes with a diameter of 8–10 mm.
This item is not returnable. View return policy
Description
The GLOBINclear-Human Kit is a rapid and robust treatment for RNA derived from human whole blood that uses a novel, non-enzymatic technology to rapidly deplete >95% of the alpha and beta globin mRNA from total RNA preparations.
Advantages of the GLOBINclear-Human Kit include:
• Improves the quality and accuracy of gene expression profiling by removing >95% of unwanted globin mRNA from whole blood total RNA to reveal biologically-relevant data
• Avoids harsh RNase H treatments that can degrade mRNA and alter expression profiles
• Produces a superior template for next-generation sequencing, RNA amplification reactions, cDNA synthesis, microarray analysis, and routine quantitative RT-PCR by using a non-enzymatic technology
Globin mRNA depletion is a vital preparatory step
Transcriptome expression profiling of whole blood samples is widely used to study physiological, pathogenic, and therapeutic responses of the transcriptome. However, up to 70% of the mRNA (by mass) in whole blood total RNA samples is composed of unwanted globin transcripts that affect the quality and accuracy of gene expression profiling in DNA microarray and sequencing technologies. Thus, globin mRNA depletion is a vital preparatory step to obtain accurate and biologically-relevant data during transcriptome analysis.
The GLOBINclear advantage
The GLOBINclear kit uses a novel hybridization technology that takes advantage of the strength of biotin/streptavidin binding, the specificity of nucleic acid hybridization, and the convenience of magnetic separations. First, globin mRNA is removed from whole blood RNA using a novel hybridization capture technology; then the remaining RNA is further purified with a rapid magnetic bead-based clean-up procedure.
As a result, GLOBINclear processed RNA provides more sensitive expression profile results than unprocessed whole blood RNA because the depletion procedure alleviates problems associated with the abundant globin mRNA found in whole blood RNA samples.
Furthermore, other globin mRNA depletion technologies rely on harsh RNase H enzymatic treatments that may compromise the expression profile by degrading mRNAs other than globin mRNA. Because the GLOBINclear kit uses a non-enzymatic technology, the resulting RNA is a superior template for next-generation sequencing, RNA amplification reactions, cDNA synthesis, microarray analysis, and routine quantitative RT-PCR.
Accessory product
The kit requires the use of a magnetic stand. The DynaMag-2 Magnetic Stand (Cat. No. 12321D) is recommended.
Alternative formats
The GLOBINclear-Human Kit is also available in an automation-friendly 96-well format for high throughput screening. For more information, please contact Technical Support.
Order Info
Shipping Condition: Approved for shipment on Wet or Dry Ice

Specifications
mRNA Depletion Kit | |
Total RNA (Globin mRNA-Depleted) | |
Total RNA (Whole Blood) | |
Microtube | |
Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting, cDNA Library Construction |
• 20 μl Capture Oligo Mix (-20°C) • 600 μl Streptavidin Magnetic Beads (4°C—do not freeze) • 1.5 ml 2X Hybridization Buffer (room temperature) • 1.5 ml Streptavidin Bead Buffer (room temperature) • 1.75 Nuclease-free Water (-20°C, 4°C, or room temperature) • 200 μl RNA Binding Beads (4°C—do not freeze) • 5 ml RNA Wash Solution Concentrate (4°C or room temperature) • 1 ml Elution Buffer (4°C or room temperature) • 80 μl RNA Bead Buffer (room temperature) • 4 ml RNA Binding Buffer Concentrate (room temperature) • 100 1.5-ml Non-stick Tubes (room temperature) All components shipped on wet ice except for Capture Oligo Mix (dry ice). |
|
High-throughput Compatible | |
Human | |
Ambion™, GLOBINclear™ | |
20 Preps |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.