missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ambion™ In Vivo Pre-Designed siRNA
Ambion™ siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.
Brand: Invitrogen™ 4457304
2231.10 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
These siRNAs are guaranteed-to-silence based on their proven design.
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
Specifications
Store in freezer (-5°C to -30°C). | |
siRNA | |
siRNA | |
1 μmol |
Ambion In Vivo Pre-Designed SIRNA HPLC, 1 umol | |
Ambion™ | |
HPLC |
For Research Use Only. Not for use in diagnostic procedures.