missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ambion™ In Vivo Pre-Designed siRNA
Ambion™ siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.
£2010.00 - £3385.00
Specifications
Content And Storage | Store in freezer (-5°C to -30°C). |
---|---|
RNAi Type | siRNA |
Product Line | Ambion™ |
Product Type | siRNA |
Purity | HPLC |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15501114
|
Invitrogen™
4457301 |
Ambion In Vivo Custom SIRNA HPLC, 250 nmol |
£2010.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15511114
|
Invitrogen™
4457303 |
Ambion In Vivo Custom SIRNA HPLC 1 umol |
£3385.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15521114
|
Invitrogen™
4457304 |
Ambion In Vivo Pre-Designed SIRNA HPLC, 1 umol |
£3330.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
These siRNAs are guaranteed-to-silence based on their proven design.
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
Specifications
Store in freezer (-5°C to -30°C). | |
Ambion™ | |
HPLC |
siRNA | |
siRNA |
For Research Use Only. Not for use in diagnostic procedures.