Learn More
Invitrogen™ Magnetic Stand-96
Description
The stand has clear polyacryl base in standard SBS 96-well microplate footprint and uses 24 extremely strong permanent magnets for fast, easy magnetic separation in little as 30 seconds. The stand sits flat on lab bench or microplate adapter on robotic workstations.
For use with MagMAX™-96 magnetic bead-based RNA isolation kits, MagMAX™ for Stabilized Blood Tubes RNA Isolation Kits, MagMAX™ FFPE Nucleic Acid Isolation Kits, and the MessageAmp™ Premier RNA Amplification Kit
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | Magnetic Stand |
| Shipping Condition | Room Temperature |
| Product Line | Ambion |
| Quantity | 1 Ea. |
| Type | Magnetic Stand |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.