missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Proteinase K Powder

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

Brand:  Invitrogen™ AM2544

435.20 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10492645

  • £544.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

Proteinase K Remains Active:
• Over a wide pH range (optimal activity between 6.5 and 9.5)
• Under denaturing conditions (e.g. the presence of SDS or urea)
• In the presence of metal chelating agents (e.g. EDTA)
• At comparatively high temperatures (optimum digestion temperature is 65°C)

Applications: Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and Quality Testing: Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit Definition: One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

T. album
Store dry powder in desiccant at –20°C.
500 mg
Ambion™
Bottle
Dry Ice
Proteinase K
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.