Learn More
Invitrogen™ Proteinase K Powder
Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.
£152.00 - £544.00
Specifications
Host Species | T. album |
---|---|
Packaging | Bottle |
Content And Storage | Store dry powder in desiccant at –20°C. |
Shipping Condition | Dry Ice |
Product Type | Proteinase K |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10340065
|
Invitrogen™
AM2542 |
100 mg |
£152.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10492645
|
Invitrogen™
AM2544 |
500 mg |
£544.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.
Proteinase K Remains Active:
• Over a wide pH range (optimal activity between 6.5 and 9.5)
• Under denaturing conditions (e.g. the presence of SDS or urea)
• In the presence of metal chelating agents (e.g. EDTA)
• At comparatively high temperatures (optimum digestion temperature is 65°C)
Applications: Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.
Performance and Quality Testing: Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.
Unit Definition: One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.
Order Info
Shipping Condition: Dry Ice
Specifications
T. album | |
Store dry powder in desiccant at –20°C. | |
Proteinase K |
Bottle | |
Dry Ice | |
Ambion™ |
Safety and Handling
- Proteinase K Powder
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : Proteinase
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.