Learn More
Invitrogen™ PSI-CLONE™ 96-Well PCR Purification Plates
High throughput compatible
Brand: Invitrogen™ 4367834
Code : 50
Additional Details : Weight : 0.72000kg
Description
Applied Biosystems PSI-Clone™ PCR 96-Well plates are for the rapid purification of PCR reaction products using either vacuum- or centrifugation-based protocols. Both protocols are fast and result in high-yield, high-purity DNA ideal for molecular biology experiments.
DNA and RNA Purification and Analysis, DNA Extraction, PCR Product Clean-Up
Specifications
96-well Plate | |
4 Plates |
For Research Use Only. Not for use in diagnostic procedures.