missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSI-CLONE™ 96-Well PCR Purification Plates

High throughput compatible

£510.00 - £4595.00

Specifications

Format 96-well Plate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10627705
missing translation for 'viewDocuments'
Invitrogen™
4367834
4 Plates This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10166664
missing translation for 'viewDocuments'
Invitrogen™
4367835
50 Plates This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Applied Biosystems PSI-Clone™ PCR 96-Well plates are for the rapid purification of PCR reaction products using either vacuum- or centrifugation-based protocols. Both protocols are fast and result in high-yield, high-purity DNA ideal for molecular biology experiments.

DNA and RNA Purification and Analysis, DNA Extraction, PCR Product Clean-Up

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

96-well Plate
Videos
SDS
Documents

Documents

Certificates

For Research Use Only. Not for use in diagnostic procedures.