Learn More
Invitrogen™ RNaseZap™ RNase Decontamination Solution
RNaseZap™ RNase Decontamination Solution is a surface decontamination solution that destroys RNases on contact. You simply spray RNaseZap™ Solution onto the surface to be decontaminated and rinse it off with RNase-free water.
Brand:  Invitrogen™ AM9784
 View more versions of this product
Product Code. 10186804
  • £834.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Show More
Specifications
Specifications
RNase
Room Temperature
RNase Control Reagent
Store at room temperature.
4 L
Ambion™, RNaseZap
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ RNaseZap™ RNase Decontamination Solution > 4 L