Learn More
Invitrogen™ RNaseZap™ RNase Decontamination Wipes Refill
Invitrogen RNaseZap™ Wipes are RNaseZap™ in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water.
£102.00 - £235.00
Specifications
Format | Wipes |
---|---|
Content And Storage | Store at room temperature. |
Shipping Condition | Room Temperature |
Reagent Type | RNase Control Reagent |
Product Line | Ambion™, ElectroZap™ |
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
10092824
|
Invitrogen™
AM9786 |
Pop-up dispenser, 100 wipes | 100 Sheets |
£102.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10504625
|
Invitrogen™
AM9788 |
Refill, 3 x 100 wipes on rolls | 300 Sheets |
£235.00
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Invitrogen RNaseZap™ Wipes are RNaseZap™ in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water. Supplied as 100 pop-up sheets in a single container.
• Convenient towel format
• Completely removes RNase contamination from glass and plastic surfaces Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free
Works immediately on contact; no baking or autoclaving necessary The popular Invitrogen™ RNaseZap™ is now provided in a convenient wipe format. RNaseZap™ Wipes are designed to eliminate RNase contamination on work surfaces, pipettors and equipment that must be RNase-free. RNaseZap™ is a formulation of three ingredients known to be active against RNase. It effectively removes even high levels of RNase contamination that similar products cannot. With thorough rinsing, RNaseZap™ leaves no residues that are inhibitory to enzymatic reactions.
Note: The components are packaged separately. The wipes and RNaseZap™ Solution are loaded into the pop-up dispenser prior to first use.
Order Info
Shipping Condition: Room temperature
Specifications
Wipes | |
Room Temperature | |
Ambion™, ElectroZap™ |
Store at room temperature. | |
RNase Control Reagent |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.