missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseZap™ RNase Decontamination Wipes Refill

Invitrogen RNaseZap™ Wipes are RNaseZap™ in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water.

Brand:  Invitrogen™ AM9786

 View more versions of this product

Product Code. 10092824

  • £102.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Invitrogen RNaseZap™ Wipes are RNaseZap™ in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water. Supplied as 100 pop-up sheets in a single container.

• Convenient towel format
• Completely removes RNase contamination from glass and plastic surfaces Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free

Works immediately on contact; no baking or autoclaving necessary The popular Invitrogen™ RNaseZap™ is now provided in a convenient wipe format. RNaseZap™ Wipes are designed to eliminate RNase contamination on work surfaces, pipettors and equipment that must be RNase-free. RNaseZap™ is a formulation of three ingredients known to be active against RNase. It effectively removes even high levels of RNase contamination that similar products cannot. With thorough rinsing, RNaseZap™ leaves no residues that are inhibitory to enzymatic reactions.

Note: The components are packaged separately. The wipes and RNaseZap™ Solution are loaded into the pop-up dispenser prior to first use.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wipes
Store at room temperature.
100 Sheets
Ambion™, ElectroZap
Pop-up dispenser, 100 wipes
Room Temperature
RNase Control Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2788129 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM9786
Lot Number2828529 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM9786
Lot Number2901398 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM9786
Lot Number2919973 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberAM9786
Lot Number2947603 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberAM9786
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ RNaseZap™ RNase Decontamination Wipes Refill > Pop-up dispenser, 100 wipes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.