Learn More
Invitrogen™ Water-Saturated Phenol, pH 6.6
Ambion Water Saturated Phenol is supplied premixed and at pH 6.6 (pH 7.9 ±0.2, with included buffer).
Brand: Invitrogen™ AM9712
148.00 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
View more versions of this product
Description
Ambion Water Saturated Phenol is supplied premixed and at pH 6.6 (pH 7.9 ±0.2, with included buffer). Water Saturated Phenol is used in RNA and DNA extraction procedures. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality tested, saturated phenols are ready-to-use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.
Order Info
Shipping Condition: Room temperature
Specifications
Phenols | |
Bottle | |
Ambion™ | |
400 mL | |
Liquid |
Store at 4°C or –20°C. | |
6.6 | |
Molecular Biology Grade | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.