Learn More
Invitrogen™ Water-Saturated Phenol, pH 6.6
Ambion Water Saturated Phenol is supplied premixed and at pH 6.6 (pH 7.9 ±0.2, with included buffer).
£104.00 - £185.00
Specifications
Chemical Name or Material | Phenols |
---|---|
Content And Storage | Store at 4°C or –20°C. |
Packaging Type | Bottle |
pH | 6.6 |
Product Line | Ambion™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10458074
|
Invitrogen™
AM9712 |
400 mL |
£185.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10361425
|
Invitrogen™
AM9710 |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Ambion Water Saturated Phenol is supplied premixed and at pH 6.6 (pH 7.9 ±0.2, with included buffer). Water Saturated Phenol is used in RNA and DNA extraction procedures. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality tested, saturated phenols are ready-to-use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.
Order Info
Shipping Condition: Room temperature
Specifications
Phenols | |
Bottle | |
Ambion™ | |
Room Temperature |
Store at 4°C or –20°C. | |
6.6 | |
Molecular Biology Grade | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.