Learn More
Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs
Use for the most accurate labeling. Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups.
Brand:  Cytiva PA22000
 View more versions of this product
Product Code. 11758268
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
    • Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
    • For use in ELISA, immunoprecipitation, and blotting applications.
    • Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.
Order Info
Each kit contains five vials of dye. Each vial is sufficient for labeling 1mg protein or 30nmol amino groups.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For the labelling of biological compounds with Cy2 Reactive Dye
Cy2 bis-Reactive Dye Pack
Labeling 1mg Protein or 30nM Amino groups
2°C to 8°C, protected from light
Yellow
Each
Five foil packs each containing dried dye to label 1mg of Protein
896.95
Cy2 bis-Reactive Dye
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1