missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester)

Use for highly accurate labeling of amine groups. Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester) contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.

Brand:  Cytiva PA17104

 View more versions of this product

Product Code. 11565095

  • £1669.00 / 10mg
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Contains derivitized CyDyes that carry only one reactive group on each dye molecule
  • pH range: 3 to 10
  • DMSO tolerant
  • Photostable
  • Example protocols included with each dye pack

TRUSTED_SUSTAINABILITY
Specifications

Specifications

For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP)
Cy7 NHS Ester
3 to 10
Cy7
10 mg
Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.