missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester)
Use for highly accurate labeling of amine groups. Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester) contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.
£69.99 - £6900.00
Specifications
Kit Contents | Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye |
---|---|
pH Range | 3 to 10 |
Product Code | Brand | Label or Dye | Product Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Label or Dye | Product Type | Quantity | Price | Quantity & Availability | |||
10697655
|
Cytiva
PA13601 |
Cy3.5 | Cy3.5 NHS Ester | 1 mg | N/A | ||||
11505055
|
Cytiva
PA15102 |
Cy5 | Cy5 NHS Ester | 50 mg |
£6900.00
50mg |
Estimated Shipment: 24-12-2024
Log in to see stock. |
|||
11515055
|
Cytiva
PA15104 |
Cy5 | Cy5 NHS Ester | 10 mg |
£2100.00
10mg |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||
11545095
|
Cytiva
PA17105 |
Cy7 | Cy7 NHS Ester | 5 mg | N/A | ||||
10379814
|
Cytiva
PA17101 |
Cy7 | Cy7 NHS Ester | 1 mg |
£318.00
1mg |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||
11565095
|
Cytiva
PA17104 |
Cy7 | Cy7 NHS Ester | 10 mg |
£1620.00
10mg |
Estimated Shipment: 24-12-2024
Log in to see stock. |
|||
11595045
|
Cytiva
PA15100 |
Cy5 | Cy5 NHS Ester | 5 mg |
£1325.09
5mg |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||
10742067
|
Cytiva
PA15101 |
Cy5 | Cy5 NHS Ester | 1 mg |
£302.00
1mg |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||
11580744
|
Cytiva
25-9004-73 |
Cy5 | Cy5 NHS Ester | 150 μg | N/A | ||||
11505085
|
Cytiva
PA15106 |
Cy5 | Cy5 NHS Ester | 25 mg |
£3744.00
25mg |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||
Description
- Contains derivitized CyDyes that carry only one reactive group on each dye molecule
- pH range: 3 to 10
- DMSO tolerant
- Photostable
- Example protocols included with each dye pack
Specifications
Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye | |
3 to 10 |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1