missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Prime Western Blotting Detection Reagent

Enables the use of highly diluted primary and secondary antibodies without reducing sensitivity. Cytiva Lifescience™ Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.

Brand:  Cytiva RPN2236

477.80 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.



 View more versions of this product

Product Code. 12994780

  • £548.00 / Pack of 3
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Includes: Solution A (luminol solution) and Solution B (peroxide solution)
This item is not returnable. View return policy

Description

Description

p>
  • Stable signal enables multiple exposures and makes reagent suitable for large experimental series
  • Provides convenient handling time between detection and completion of experiment
  • Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging), Amersham Hybond™ P (PVDF) membranes and compatible with Amersham Hyperfilm™, Amersham Hybond- ECL (nitrocellulose) membranes
  • Also compatible with Rainbow™ Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers

Education and research

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ECL Western Blot Detection Reagent
∼1 pg
Solution A (Luminol solution, 3 × 50mL) and Solution B (Peroxide solution, 3 × 50mL)
Sensitive Quantitative or Confirmatory Western Blot
3000cm2 membrane
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ECL Prime

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360D-May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : trometamol, ethanediol, imidazole
  • UNKNOWN AQUATIC-Contains 4% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers